Assay Details
Gene Symbol: Crebrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277157.1 | NP_001264086.1 | 5 - 6 | 1661 | 88 | |
XM_008767581.1 | XP_008765803.1 | 4 - 5 | 1881 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
