Assay Details
Gene Symbol: Mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105834.1 | NP_001099304.1 | 2 - 3 | 286 | 95 | |
GenBank mRNA | FQ216878.1 | - | 1 - 2 | 78 | 95 | |
FQ225351.1 | - | 1 - 2 | 80 | 95 | ||
FQ226279.1 | - | 1 - 2 | 78 | 95 | ||
FQ229232.1 | - | 1 - 2 | 78 | 95 | ||
FQ232005.1 | - | 1 - 2 | 78 | 95 | ||
FQ233183.1 | - | 1 - 2 | 78 | 95 | ||
FQ233508.1 | - | 1 - 2 | 78 | 95 | ||
FQ233844.1 | - | 1 - 2 | 78 | 95 | ||
FQ234322.1 | - | 1 - 2 | 78 | 95 | ||
FQ234954.1 | - | 1 - 2 | 78 | 95 | ||
FQ235000.1 | - | 1 - 2 | 78 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
