Assay Details
Gene Symbol: Dgke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039341.1 | NP_001034430.1 | 1 - 2 | 625 | 66 | |
XM_006247070.2 | XP_006247132.1 | 1 - 2 | 860 | 66 | ||
XM_006247071.2 | XP_006247133.1 | 1 - 2 | 860 | 66 | ||
GenBank mRNA | BC110049.1 | - | 1 - 2 | 625 | 7316890 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
