Assay Details
Gene Symbol: Senp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013116.2 | NP_001013134.2 | 1 - 2 | 162 | 90 | |
XM_006246703.2 | XP_006246765.1 | 1 - 2 | 211 | 90 | ||
GenBank mRNA | BF407260.1 | - | 1 - 2 | 162 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001013116.2, XM_006246703.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
