Assay Details
Gene Symbol: Mmp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079888.1 | NP_001073357.1 | 7 - 8 | 1173 | 75 | |
GenBank mRNA | EF121988.1 | - | 7 - 8 | 1173 | 75 | |
EF121989.1 | - | 7 - 8 | 1173 | 75 |
Gene Symbol: LOC103694864 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773811.1 | XP_008772033.1 | 7 - 8 | 1364 | 75 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
