Assay Details
Gene Symbol: Afmid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111366.1 | NP_001104836.1 | 4 - 5 | 333 | 103 | |
XM_006247823.2 | XP_006247885.1 | 4 - 5 | 349 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
