Assay Details
Gene Symbol: Timd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246172.2 | XP_006246234.2 | 6 - 7 | 926 | 81 | |
XM_006246173.2 | XP_006246235.2 | 6 - 7 | 926 | 81 | ||
XM_008767718.1 | XP_008765940.1 | 6 - 7 | 990 | 81 | ||
XM_008767720.1 | XP_008765942.1 | 6 - 7 | 989 | 81 | ||
XM_008767721.1 | XP_008765943.1 | 6 - 7 | 989 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
