Assay Details
Gene Symbol: Timd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246173.2 | XP_006246235.2 | 8 - 9 | 1052 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Choose the brand aligned with your industry so we can best serve your needs.
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Timd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246173.2 | XP_006246235.2 | 8 - 9 | 1052 | 81 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027288 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027288 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P027288 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20891530 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365284 | - | - |