Assay Details
Gene Symbol: Pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768473.1 | XP_008766695.1 | 1 - 2 | 65 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768473.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
