Assay Details
Gene Symbol: Pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105857.2 | NP_001099327.1 | 2 - 3 | 137 | 73 | |
XM_008768472.1 | XP_008766694.1 | 3 - 4 | 334 | 73 | ||
XM_008768473.1 | XP_008766695.1 | 3 - 4 | 268 | 73 | ||
XM_008768474.1 | XP_008766696.1 | 3 - 4 | 258 | 73 | ||
XM_008768475.1 | XP_008766697.1 | 3 - 4 | 334 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
