Assay Details
Gene Symbol: Trim41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134737.1 | NP_001128209.1 | 5 - 6 | 1991 | 60 | |
GenBank mRNA | BC168206.1 | - | 5 - 6 | 1036 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
