Assay Details
Gene Symbol: Poldip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105816.1 | NP_001099286.1 | 2 - 3 | 185 | 80 | |
GenBank mRNA | FJ515740.1 | - | 355 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105816.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
