Assay Details
Gene Symbol: Shmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047842.1 | NP_001041307.1 | 7 - 8 | 952 | 96 | |
XM_006246469.2 | XP_006246531.1 | 4 - 5 | 735 | 96 | ||
GenBank mRNA | AY383687.1 | - | 7 - 8 | 952 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
