Assay Details
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007607.1 | NP_001007608.1 | 4 - 5 | 992 | 76 | |
XM_006245942.2 | XP_006246004.1 | 4 - 5 | 1071 | 76 | ||
XM_008767559.1 | XP_008765781.1 | 4 - 5 | 1011 | 76 | ||
XM_008767560.1 | XP_008765782.1 | 4 - 5 | 1038 | 76 | ||
GenBank mRNA | BC081875.1 | - | 4 - 5 | 992 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
