Assay Details
Gene Symbol: Arhgdia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007005.1 | NP_001007006.1 | 2 - 3 | 267 | 62 | |
XM_006247897.2 | XP_006247959.1 | 2 - 3 | 401 | 62 | ||
XM_006247898.2 | XP_006247960.1 | 2 - 3 | 350 | 62 | ||
GenBank mRNA | BC083817.1 | - | 2 - 3 | 267 | 7110945 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
