Assay Details
Gene Symbol: Smyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105810.1 | NP_001099280.1 | 1 - 2 | 264 | 62 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001105810.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
