Assay Details
Gene Symbol: Smyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105810.1 | NP_001099280.1 | 3 - 4 | 552 | 84 | |
XM_006246674.2 | XP_006246736.1 | 3 - 4 | 456 | 84 | ||
XM_008767962.1 | XP_008766184.1 | 2 - 3 | 681 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
