Assay Details
Gene Symbol: Smyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105810.1 | NP_001099280.1 | 9 - 10 | 2398 | 80 | |
XM_006246674.2 | XP_006246736.1 | 9 - 10 | 2302 | 80 | ||
XM_008767962.1 | XP_008766184.1 | 8 - 9 | 2527 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
