Assay Details
Gene Symbol: Mrpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105781.1 | NP_001099251.1 | 1 - 2 | 56 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105781.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
