Assay Details
Gene Symbol: Wipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191825.1 | NP_001178754.1 | 4 - 5 | 975 | 142 | |
XM_006247353.2 | XP_006247415.1 | 5 - 6 | 1201 | 142 | ||
XM_008768080.1 | XP_008766302.1 | 5 - 6 | 1202 | 142 | ||
XM_008768081.1 | XP_008766303.1 | 5 - 6 | 1240 | 142 | ||
XM_008768082.1 | XP_008766304.1 | 5 - 6 | 2885 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
