Assay Details
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213625.1 | NP_998790.1 | 1 - 2 | 77 | 164 | |
XM_006247311.2 | XP_006247373.1 | 2 - 3 | 438 | 164 | ||
GenBank mRNA | AB178470.1 | - | 1 - 2 | 77 | 164 | |
FQ212948.1 | - | 1 - 2 | 108 | 164 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
