Assay Details
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007675.1 | NP_001007676.1 | 1 - 2 | 257 | 85 | |
GenBank mRNA | BC083720.1 | - | 1 - 2 | 257 | 85 | |
FQ209955.1 | - | 1 - 2 | 358 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC083720.1, NM_001007675.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
