Assay Details
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007675.1 | NP_001007676.1 | 2 - 3 | 423 | 65 | |
XM_006247800.2 | XP_006247862.1 | 2 - 3 | 949 | 65 | ||
XM_006247801.2 | XP_006247863.1 | 2 - 3 | 564 | 65 | ||
XM_006247802.2 | XP_006247864.1 | 2 - 3 | 1021 | 65 | ||
XM_008768374.1 | XP_008766596.1 | 2 - 3 | 625 | 65 | ||
GenBank mRNA | BC083720.1 | - | 2 - 3 | 423 | 65 | |
FQ209955.1 | - | 2 - 3 | 524 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
