Assay Details
Gene Symbol: Stac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108834.1 | NP_001102304.1 | 3 - 4 | 992 | 90 | |
XM_006247383.2 | XP_006247445.1 | 5 - 6 | 1072 | 90 | ||
GenBank mRNA | AM903074.1 | - | 5 - 6 | 706 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
