Assay Details
Gene Symbol: Stac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108834.1 | NP_001102304.1 | 2 - 3 | 882 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
