Assay Details
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012497.1 | NP_036629.1 | 2 - 3 | 220 | 70 | |
GenBank mRNA | BC059154.1 | - | 2 - 3 | 195 | 70 | |
BC099749.1 | - | 2 - 3 | 206 | 70 | ||
FQ213506.1 | - | 2 - 3 | 223 | 70 | ||
FQ214420.1 | - | 2 - 3 | 246 | 70 | ||
FQ228893.1 | - | 2 - 3 | 220 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC059154.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
