Assay Details
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012751.1 | NP_036883.1 | 2 - 3 | 289 | 80 | |
XM_006246596.2 | XP_006246658.1 | 3 - 4 | 262 | 80 | ||
GenBank mRNA | BC085757.1 | - | 2 - 3 | 334 | 80 | |
D28561.1 | - | 2 - 3 | 289 | 80 | ||
FQ216450.1 | - | 3 - 4 | 228 | 80 | ||
FQ232657.1 | - | 2 - 3 | 348 | 80 | ||
FQ233419.1 | - | 2 - 3 | 324 | 80 | ||
J04524.1 | - | 2 - 3 | 271 | 80 | ||
KJ696745.1 | - | 2 - 3 | 174 | 80 | ||
M25482.1 | - | 2 - 3 | 254 | 80 | ||
X14771.1 | - | 2 - 3 | 302 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
