Assay Details
Gene Symbol: Lcorl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770224.1 | XP_008768446.1 | 5 - 6 | 566 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
