Assay Details
Gene Symbol: Eda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257110.2 | XP_006257172.1 | 5 - 6 | 937 | 62 | |
XM_006257111.2 | XP_006257173.1 | 5 - 6 | 937 | 62 | ||
XM_006257112.2 | XP_006257174.1 | 5 - 6 | 937 | 62 | ||
XM_008773301.1 | XP_008771523.1 | 5 - 6 | 937 | 62 | ||
XM_228582.8 | XP_228582.6 | 5 - 6 | 937 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
