Assay Details
Gene Symbol: Mbtps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035007.1 | NP_001030179.1 | 8 - 9 | 1155 | 85 | |
XM_006256944.2 | XP_006257006.1 | 6 - 7 | 844 | 85 | ||
GenBank mRNA | BC097391.1 | - | 1117 | 7453173 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -




Gene Ontology Categories:
MicroArray Probes:
