Assay Details
Gene Symbol: Il2rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080889.1 | NP_543165.1 | 6 - 7 | 870 | 134 | |
GenBank mRNA | AY325195.1 | - | 698 | 134 | ||
BC079343.1 | - | 6 - 7 | 870 | 7106866 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
