Assay Details
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270598.1 | NP_001257527.1 | 3 - 4 | 368 | 139 | |
NM_001270599.1 | NP_001257528.1 | 3 - 4 | 368 | 139 | ||
GenBank mRNA | FQ222567.1 | - | 3 - 4 | 136 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270599.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
