Assay Details
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142363.1 | NP_001135835.1 | 4 - 5 | 628 | 94 | |
NR_024535.1 | 3 - 4 | 528 | 94 | |||
XM_006248558.2 | XP_006248620.1 | 4 - 5 | 646 | 94 | ||
GenBank mRNA | BC088475.1 | - | 3 - 4 | 511 | 94 | |
FM075565.1 | - | 3 - 4 | 440 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
