Assay Details
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142363.1 | NP_001135835.1 | 6 - 7 | 952 | 65 | |
NR_024535.1 | 5 - 6 | 852 | 65 | |||
GenBank mRNA | BC088475.1 | - | 5 - 6 | 835 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
