Assay Details
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142363.1 | NP_001135835.1 | 8 - 9 | 1171 | 126 | |
NR_024535.1 | 7 - 8 | 1071 | 126 | |||
XM_006248558.2 | XP_006248620.1 | 7 - 8 | 1130 | 126 | ||
GenBank mRNA | BC088475.1 | - | 7 - 8 | 1054 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
