Assay Details
Gene Symbol: Ndfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108390.2 | NP_001101860.2 | 6 - 7 | 1011 | 108 | |
XM_006252429.1 | XP_006252491.1 | 7 - 8 | 956 | 108 | ||
GenBank mRNA | DY565248.1 | - | 5 - 6 | 573 | 108 | |
FM044778.1 | - | 1 - 2 | 60 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
