Assay Details
Gene Symbol: Ndfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108390.2 | NP_001101860.2 | 3 - 4 | 728 | 94 | |
XM_006252429.1 | XP_006252491.1 | 4 - 5 | 673 | 94 | ||
GenBank mRNA | CO398212.1 | - | 3 - 4 | 511 | 94 | |
DY565248.1 | - | 2 - 3 | 290 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
