Assay Details
Gene Symbol: Lcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012044.1 | NP_001012044.1 | 2 - 3 | 108 | 89 | |
XM_006252318.2 | XP_006252380.1 | 2 - 3 | 283 | 89 | ||
XM_006252319.2 | XP_006252381.1 | 4 - 5 | 482 | 89 | ||
XM_006252320.2 | XP_006252382.1 | 4 - 5 | 530 | 89 | ||
XM_006252322.2 | XP_006252384.1 | 2 - 3 | 1689 | 89 | ||
GenBank mRNA | BC083855.1 | - | 2 - 3 | 108 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
