Assay Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100685.1 | NP_001094155.1 | 1 - 2 | 76 | 87 | |
XM_006252508.2 | XP_006252570.1 | 1 - 2 | 94 | 87 | ||
GenBank mRNA | BC088243.1 | - | 1 - 2 | 56 | 87 | |
FM063772.1 | - | 1 - 2 | 76 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
