Assay Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100685.1 | NP_001094155.1 | 6 - 7 | 809 | 150 | |
XM_006252508.2 | XP_006252570.1 | 5 - 6 | 638 | 150 | ||
GenBank mRNA | BC088243.1 | - | 6 - 7 | 789 | 150 | |
FQ214276.1 | - | 5 - 6 | 594 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
