Assay Details
Gene Symbol: Eif2s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199380.1 | NP_955412.1 | 1 - 2 | 88 | 117 | |
GenBank mRNA | BC062402.1 | - | 1 - 2 | 88 | 117 | |
FQ215337.1 | - | 1 - 2 | 133 | 117 | ||
FQ215587.1 | - | 1 - 2 | 29 | 117 | ||
FQ222905.1 | - | 1 - 2 | 133 | 117 | ||
FQ234002.1 | - | 1 - 2 | 118 | 117 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC062402.1, NM_199380.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
