Assay Details
Gene Symbol: Ccm2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762395.1 | XP_008760617.1 | 7 - 8 | 1879 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
