Assay Details
Gene Symbol: Sdcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025692.1 | NP_001020863.1 | 1 - 2 | 62 | 70 | |
XM_008762326.1 | XP_008760548.1 | 2 - 3 | 1043 | 70 | ||
GenBank mRNA | BC099095.1 | - | 1 - 2 | 62 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC099095.1, NM_001025692.1, XM_008762326.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
