Assay Details
Gene Symbol: Sdcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025692.1 | NP_001020863.1 | 5 - 6 | 467 | 125 | |
XM_008762325.1 | XP_008760547.1 | 3 - 4 | 2443 | 125 | ||
XM_008762326.1 | XP_008760548.1 | 6 - 7 | 1448 | 125 | ||
GenBank mRNA | BC099095.1 | - | 5 - 6 | 467 | 7459023 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
