Assay Details
Gene Symbol: Pofut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002278.1 | NP_001002278.1 | 4 - 5 | 559 | 59 | |
XM_006235290.2 | XP_006235352.2 | 4 - 5 | 703 | 59 | ||
GenBank mRNA | AJ781499.1 | - | 4 - 5 | 559 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
