Assay Details
Gene Symbol: Pofut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002278.1 | NP_001002278.1 | 5 - 6 | 759 | 66 | |
XM_006235290.2 | XP_006235352.2 | 5 - 6 | 903 | 66 | ||
GenBank mRNA | AJ781499.1 | - | 5 - 6 | 759 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
