Assay Details
Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106529.1 | NP_001099999.1 | 6 - 7 | 1998 | 162 | |
XM_006235287.2 | XP_006235349.1 | 6 - 7 | 2419 | 162 | ||
XM_006235288.1 | XP_006235350.1 | 6 - 7 | 2035 | 162 | ||
XM_006235289.2 | XP_006235351.1 | 6 - 7 | 2215 | 162 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
