Assay Details
Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106529.1 | NP_001099999.1 | 7 - 8 | 2115 | 77 | |
XM_006235287.2 | XP_006235349.1 | 7 - 8 | 2536 | 77 | ||
XM_006235288.1 | XP_006235350.1 | 7 - 8 | 2152 | 77 | ||
XM_006235289.2 | XP_006235351.1 | 7 - 8 | 2332 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
