Assay Details
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031068.1 | NP_112330.1 | 7 - 8 | 774 | 115 | |
GenBank mRNA | AB292042.1 | - | 7 - 8 | 698 | 115 | |
AB292043.1 | - | 7 - 8 | 687 | 115 | ||
BC098014.1 | - | 7 - 8 | 774 | 115 | ||
FQ212256.1 | - | 7 - 8 | 976 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
