Assay Details
Gene Symbol: Acvr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031571.2 | NP_113759.1 | 4 - 5 | 553 | 78 | |
GenBank mRNA | L10639.1 | - | 4 - 5 | 536 | 78 | |
S48190.1 | - | 4 - 5 | 553 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
